Learn More
Medchemexpress LLC SDMA-d6 (Synonyms: Symmetric dimethylarginine-d6; NG,NG'-Dimethyl-L-arginine-d6), 1mg
SDP
Catalog No. NC3242638
Encompass_Preferred
SDMA-d6 is the deuterium labeled SDMA. SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity[1][2].
Catalog No. NC3242638 Supplier Medchemexpress LLC Supplier No. HY101410S1MG
SDMA-d6 is the deuterium labeled SDMA. SDMA (Symmetric dimethylarginine) is an endogenous inhibitor of nitric oxide (NO) synthase activity[1][2].