Learn More
Enzo Life Sciences SDMA ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ALX850331KI01
Encompass_Preferred
The SDMA ELISA kit is a complete assay for the quantitative determination of symmetric dimethylarginine (SDMA) levels. This kit is to be used with EDTA-plasma and serum from human origin. The SDMA ELISA has no cross-reactivity with ADMA, NMMA or L-arginine. Alternative name: NG, N’G-Dimethyl-L-arginine ELISA kit. Assay Time: 15 to 20 hours + 1 hour. Applications: ELISA . Species reactivity: Human .
Catalog No. 50-201-0164