Learn More
Supply Solutions Sodium dodecyl sulfate, 151-21-3, MFCD00036175, 1kg
Supplier:  Supply Solutions SIGMA L37711KG
Encompass_Preferred
Linear Formula CH3(CH2)11OSO3Na, ?98.5% (GC), Molecular Weight 288.38, BioReagent, suitable for electrophoresis, for molecular biology, Synonym: Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate
Catalog No. NC0924094