Learn More
Biotang Inc SDS (10%) 250 ML, BM-230A, 10% SDS. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Supplier:  Biotang Inc BTBB539
Encompass_Preferred
SDS (10%) 250 ML, BM-230A, 10% SDS. Prepared in 18.2 megohms water and filtered through 1-micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-155-861