Learn More
Boston Bioproducts Inc Sodium Dodecyl Sulfate, SDS (20%) - 250ml
Catalog No. NC9380630
Encompass
Sodium Dodecyl Sulfate (SDS), also known as Sodium Lauryl Sulfate (SLS), is an anionic surfactant consisting of a 12-carbon tail attached to a sulfate group, giving it amphiphilic properties. This means that it has both hydrophobic and hydrophillic parts, which enables the formation of micelles in solution capable of solubilizing hydrophobic targets in many molecular biology applications including nucleic acids extraction and protein electrophoresis.
Catalog No. NC9380630 Supplier Boston Bioproducts Inc Supplier No. BM230250ML
Sodium Dodecyl Sulfate (SDS), also known as Sodium Lauryl Sulfate (SLS), is an anionic surfactant consisting of a 12-carbon tail attached to a sulfate group, giving it amphiphilic properties. This means that it has both hydrophobic and hydrophillic parts, which enables the formation of micelles in solution capable of solubilizing hydrophobic targets in many molecular biology applications including nucleic acids extraction and protein electrophoresis.