Learn More
Quality Biological Inc SDS 20%, 500 mL
SDP
Supplier:  Quality Biological Inc 351066101
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. ¿ SDS is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. ¿ SDS is also a powerful protein denaturant. ¿ Popular anionic detergent for a variety of protein methods ¿ Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE) ¿ Common component of cell lysis buffers ¿ Storage Conditions: 25¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase, and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7401