Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises SDS Binder

Catalog No. 18003015
Encompass
Encompass_Preferred
Change view
Click to view available options
Height (English):
12 in.
12.5 in.
Width (English):
1.5 in.
2.5 in.
3 in.
Diameter (English) Ring:
1.5 in.
2.5 in.
3 in.
Languages:
English
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English) Diameter (English) Ring Languages
18-003-015 12.5 in. 3 in. 3 in. English
18-003-013 12 in. 1.5 in. 1.5 in. English
18-003-014 12 in. 2.5 in. 2.5 in. English
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-003-015 Supplier ZING™ Enterprises Supplier No. 6032
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Each binder contains a heavy-duty ring assembly, available in 3 sizes.

ZING SDS Binder prominently stores important safety and training documents to comply with the new GHS/Hazard Communication requirements. Binder is constructed from high quality 100% post-consumer poly materials that reinforce sustainability initiatives as well as compliance. Binder is highly visible with its bright red/white colors and is extremely durable, chemical resistant and easy to clean.

Different sizes include – 1.5 in. (up to 300 sheets), 2.5 in. (up to 500 sheets), and 3 in. (up to 600 sheets). Binders are available in English and bilingual English/Spanish versions.

Specifications

Description Floor standing
Product Type Hazard Communication
Diameter (English) Ring 3 in.
Width (English) 3 in.
Height (English) 12.5 in.
Quantity 1
Material Plastic
Color Red, White
Languages English
For Use With (Application) SDS Management
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.