Learn More
MilliporeSigma SDS LYSIS BUFFER
Catalog No. NC9118931
Encompass
SDS Lysis Buffer, Category: Nuclear Function, Pack size: 10mL, Tested Application: Chromatin Immunoprecipitation assays, Formulation: 10mL of 1% SDS, 10mM EDTA and 50mM Tris, 8.1pH, Liquid
Catalog No. NC9118931 Supplier MilliporeSigma Supplier No. 20163
SDS Lysis Buffer, Category: Nuclear Function, Pack size: 10mL, Tested Application: Chromatin Immunoprecipitation assays, Formulation: 10mL of 1% SDS, 10mM EDTA and 50mM Tris, 8.1pH, Liquid