Learn More
Alkali Scientific SDS, Micro Pellets [Sodium dodecylsulfate][Lauryl sulfate], 5 Kg
SDP
Catalog No. 5000661144
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS) micro pellets, available in 5kg bulk form, are an essential anionic detergent used for large-scale protein denaturation and analysis. SDS is a powerful tool in SDS-PAGE, where it disrupts the tertiary and quaternary structures of proteins, allowing them to unfold and migrate based on size. The high concentration of SDS in these micro pellets ensures that large volumes of solution can be quickly prepared for protein analysis, making it ideal for high-throughput laboratories. This product is also used for membrane protein solubilization and in any application where protein denaturation and separation by size are required.
Catalog No. 50-006-61144 Supplier Alkali Scientific Supplier No. SDS013CP
Sodium Dodecyl Sulfate (SDS) micro pellets, available in 5kg bulk form, are an essential anionic detergent used for large-scale protein denaturation and analysis. SDS is a powerful tool in SDS-PAGE, where it disrupts the tertiary and quaternary structures of proteins, allowing them to unfold and migrate based on size. The high concentration of SDS in these micro pellets ensures that large volumes of solution can be quickly prepared for protein analysis, making it ideal for high-throughput laboratories. This product is also used for membrane protein solubilization and in any application where protein denaturation and separation by size are required.