Learn More
Alkali Scientific SDS, Micro Pellets [Sodium dodecylsulfate][Lauryl sulfate], 500 G
SDP
Catalog No. 5000661591
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS) micro pellets in a 500g size are designed for protein denaturation, particularly in SDS-PAGE analysis. SDS is an essential anionic detergent that disrupts non-covalent bonds in proteins, ensuring that they unfold and migrate according to their size during electrophoresis. These pellets offer the advantage of quick dissolution and minimal residue, providing an efficient means of preparing SDS solutions for protein analysis. Laboratories working with membrane proteins, protein extraction, and solubilization in biochemical studies will find these SDS pellets particularly useful for consistent and reproducible results.
Catalog No. 50-006-61591 Supplier Alkali Scientific Supplier No. SDS012CP
Sodium Dodecyl Sulfate (SDS) micro pellets in a 500g size are designed for protein denaturation, particularly in SDS-PAGE analysis. SDS is an essential anionic detergent that disrupts non-covalent bonds in proteins, ensuring that they unfold and migrate according to their size during electrophoresis. These pellets offer the advantage of quick dissolution and minimal residue, providing an efficient means of preparing SDS solutions for protein analysis. Laboratories working with membrane proteins, protein extraction, and solubilization in biochemical studies will find these SDS pellets particularly useful for consistent and reproducible results.