Learn More
BOSTER BIOLOGICAL TECHNOLOGY LTD CO SDS-PAGE PROT LOAD BUFF 5X-3ML
SDP
Catalog No. NC1982640
Encompass
Boster’s SDS PAGE Sample Buffer 5X (Reducing) is the most commonly used sample buffer for Sodium Dodecyl Sulfate - Polyacrylamide Gel Electrophoresis (SDS-PAGE) of denatured proteins in the Laemmli SDS-PAGE system.This buffer is very important in the preparation of protein samples and loading them onto a gel. Boster protein loading buffer is a 5X reducing hence it requires less sample dilution but yet accommodating more proteins load in a well. BOSTER IMMUNOLEADER mfg# AR-1112
Catalog No. NC1982640 Supplier BOSTER BIOLOGICAL TECHNOLOGY LTD CO Supplier No. AR11123ML
Boster’s SDS PAGE Sample Buffer 5X (Reducing) is the most commonly used sample buffer for Sodium Dodecyl Sulfate - Polyacrylamide Gel Electrophoresis (SDS-PAGE) of denatured proteins in the Laemmli SDS-PAGE system.This buffer is very important in the preparation of protein samples and loading them onto a gel. Boster protein loading buffer is a 5X reducing hence it requires less sample dilution but yet accommodating more proteins load in a well. BOSTER IMMUNOLEADER mfg# AR-1112