Learn More
Research Products International Corp SDS [Sodium dodecylsulfate] [Lauryl sulfate], Powder, 100 Grams
SDP
Catalog No. 50213393
Encompass_Preferred
Ionic detergent for denaturing proteins prior to gel electrophoresis.
Catalog No. 50-213-393 Supplier Research Products International Corp Supplier No. L22010100.0
Ionic detergent for denaturing proteins prior to gel electrophoresis.