Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc SDS Protein Gel Loading Solution (2X), 10 mL
SDP

Catalog No. 507517251
Encompass_Preferred

SDS Protein Gel Loading Solution (2X) is formulated at the traditional 2X stocks for standard volume of sample to be loaded per well. Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques.

Applications: SDS Protein Gel Loading Solution, 2X is a convenient and ready-to-use solution for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7251 Supplier Quality Biological Inc Supplier No. 351082661EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SDS Protein Gel Loading Solution (2X) is formulated at the traditional 2X stocks for standard volume of sample to be loaded per well. Simply load the gel sample wells and visualize progress of the electrophoresis run by monitoring the location of the blue dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques.

Applications: SDS Protein Gel Loading Solution, 2X is a convenient and ready-to-use solution for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Endotoxin and Mycoplasma Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.