Learn More
Boston Bioproducts Inc Laemmli SDS-Sample Buffer (4X, Reducing) - 25ml
Catalog No. NC9176164
Encompass
The 4X laemmli sample buffer is a critical component in the preparation of protein samples for SDS-PAGE, ensuring the resolution of protein bands. Its composition typically includes Tris-HCl, glycerol, SDS, and a tracking dye like Bromophenol Blue. In the reducing Laemmli sample buffer, a reducing agent such as β-mercaptoethanol or dithiothreitol (DTT) is added to break disulfide bonds within protein subunits, allowing them separate by size in electrophoresis.
Catalog No. NC9176164 Supplier Boston Bioproducts Inc Supplier No. BP110R25ML
The 4X laemmli sample buffer is a critical component in the preparation of protein samples for SDS-PAGE, ensuring the resolution of protein bands. Its composition typically includes Tris-HCl, glycerol, SDS, and a tracking dye like Bromophenol Blue. In the reducing Laemmli sample buffer, a reducing agent such as β-mercaptoethanol or dithiothreitol (DTT) is added to break disulfide bonds within protein subunits, allowing them separate by size in electrophoresis.