Learn More
AB Sciex LLC SDS Sample Buffer, 50 mL, 100 mM Tris-HCl, pH 9, 1% SDS, for Capillary Electrophoresis (CE), research use only
Catalog No. NC3405953
Encompass
SDS Sample Buffer, 50 mL, 100 mM Tris-HCl, pH 9, 1% SDS, for Capillary Electrophoresis (CE), research use only
Catalog No. NC3405953 Supplier AB Sciex LLC Supplier No. 5314719
SDS Sample Buffer, 50 mL, 100 mM Tris-HCl, pH 9, 1% SDS, for Capillary Electrophoresis (CE), research use only