Learn More
Bioworld SDS Sample Loading Buffer (4X), 250 mL
Catalog No. 50198643
Encompass_Preferred
Product DescriptionSDS sample loading buffer has a 4X concentration. A 1X concentration contains 50 mM Tris-HCl at a pH value of 6.8, 2% SDS, 10% glycerol, 1% β-mercaptoethanol, 12.5 mM EDTA, and 0.02% bromophenol blue. Use 1X for loading samples onto SDS-polyacrylamide gels.
Catalog No. 50-198-643 Supplier Bioworld Supplier No. 214200183
Product DescriptionSDS sample loading buffer has a 4X concentration. A 1X concentration contains 50 mM Tris-HCl at a pH value of 6.8, 2% SDS, 10% glycerol, 1% β-mercaptoethanol, 12.5 mM EDTA, and 0.02% bromophenol blue. Use 1X for loading samples onto SDS-polyacrylamide gels.