Learn More
Alkali Scientific SDS [Sodium dodecylsulfate] [Lauryl sulfate], Powder, 1 Kg
SDP
Catalog No. 5000661294
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS), Powder, 1kg, is a high-quality detergent used extensively in molecular biology and biochemistry. SDS is commonly used for protein denaturation, breaking down the secondary, tertiary, and quaternary structures of proteins, ensuring accurate analysis in SDS-PAGE (Polymerase Gel Electrophoresis). The powder form offers flexibility in preparing customized concentrations for various applications. It is essential for protein analysis, purification, and characterization. The laboratory-grade quality ensures high purity, enabling consistent and reproducible results in experiments. SDS is a critical reagent for proteomics, protein research, and cell lysis protocols.
Catalog No. 50-006-61294 Supplier Alkali Scientific Supplier No. SDS001CP
Sodium Dodecyl Sulfate (SDS), Powder, 1kg, is a high-quality detergent used extensively in molecular biology and biochemistry. SDS is commonly used for protein denaturation, breaking down the secondary, tertiary, and quaternary structures of proteins, ensuring accurate analysis in SDS-PAGE (Polymerase Gel Electrophoresis). The powder form offers flexibility in preparing customized concentrations for various applications. It is essential for protein analysis, purification, and characterization. The laboratory-grade quality ensures high purity, enabling consistent and reproducible results in experiments. SDS is a critical reagent for proteomics, protein research, and cell lysis protocols.