Learn More
Alkali Scientific SDS [Sodium dodecylsulfate] [Lauryl sulfate], Powder, 2.5 Kg
SDP
Catalog No. 5000661592
Encompass_Preferred
Sodium Dodecyl Sulfate (SDS), Powder, 2.5kg, is a high-quality detergent used for protein denaturation and analysis in various molecular biology and biochemistry applications. SDS is commonly used in SDS-PAGE for separating proteins based on their molecular weight and is essential for protein purification, characterization, and analysis. The 2.5kg bulk package is ideal for labs with high throughput, providing a cost-effective solution for large-scale experiments. SDS is also used for nucleic acid extraction, cell lysis, and protein solubilization. The laboratory-grade SDS ensures high purity and performance, enabling consistent and reliable results in all protein-based assays.
Catalog No. 50-006-61592 Supplier Alkali Scientific Supplier No. SDS003CP
Sodium Dodecyl Sulfate (SDS), Powder, 2.5kg, is a high-quality detergent used for protein denaturation and analysis in various molecular biology and biochemistry applications. SDS is commonly used in SDS-PAGE for separating proteins based on their molecular weight and is essential for protein purification, characterization, and analysis. The 2.5kg bulk package is ideal for labs with high throughput, providing a cost-effective solution for large-scale experiments. SDS is also used for nucleic acid extraction, cell lysis, and protein solubilization. The laboratory-grade SDS ensures high purity and performance, enabling consistent and reliable results in all protein-based assays.