Learn More
Crescent Chemical Co Inc SDS SOLUTION 20%
SDP
Catalog No. 50247548
Encompass_Preferred
SDS Solution, 20 % SDS (cat. no. 20770): 200 g/L Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 20767.02
Catalog No. 50-247-548 Supplier Crescent Chemical Co Inc Supplier No. SE20767.02
Add to Cart
Add to Cart
SDS Solution, 20 % SDS (cat. no. 20770): 200 g/L Size - 500ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 20767.02