Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioland Scientific LLC SDS SOLUTION (20% W/V, 1 L)
SDP

Catalog No. NC1362628
Encompass_Preferred

SDS Solution (20% W/V, 1 L)

Catalog No. NC1362628 Supplier Bioland Scientific LLC Supplier No. SDS0202
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SDS Solution (20% W/V, 1 L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.