Learn More
Crescent Chemical Co Inc SDS SOLUTION 20%
SDP
Supplier:  Crescent Chemical Co Inc SE20767.03
Encompass_Preferred
SDS Solution, 20 % SDS (cat. no. 20770): 200 g/L Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 20767.03
Catalog No. 50-247-549