Learn More
Cayman Chemical SDuloxetInhydrochLRIde 100mg
SDP
Catalog No. 502952785
Encompass_Preferred
A serotonin and norepinephrine reuptake inhibitor (Kis = 4.6 and 15.6 nM, respectively); also inhibits dopamine reuptake (Ki = 369 nM); suppresses spontaneous firing activity in vivo in the dorsal raphe and locus coeruleus (ED50s = 99 and 475 UG/kg, respectively); has antidepressant-like activity in mice in the forced swim test at 16 and 32 mg/kg
Catalog No. 50-295-2785 Supplier Cayman Chemical Supplier No. 14317100 MG
A serotonin and norepinephrine reuptake inhibitor (Kis = 4.6 and 15.6 nM, respectively); also inhibits dopamine reuptake (Ki = 369 nM); suppresses spontaneous firing activity in vivo in the dorsal raphe and locus coeruleus (ED50s = 99 and 475 UG/kg, respectively); has antidepressant-like activity in mice in the forced swim test at 16 and 32 mg/kg