Learn More
Medchemexpress LLC HY-13059B 10mg Medchemexpress, SDZ 220-581 (hydrochloride) CAS:179411-93-9 Purity:>98%
SDP
Catalog No. 501932651
Encompass_Preferred
Medchemexpress, HY-13059B 10mg SDZ 220-581 (hydrochloride) CAS:179411-93-9 SDZ 220-581 hydrochloride is an oral bioavailability, potent, competitive NMDA receptor antagonist with pK i value of 7.7 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2651 Supplier Medchemexpress LLC Supplier No. HY13059B10MG
Medchemexpress, HY-13059B 10mg SDZ 220-581 (hydrochloride) CAS:179411-93-9 SDZ 220-581 hydrochloride is an oral bioavailability, potent, competitive NMDA receptor antagonist with pK i value of 7.7 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.