Learn More
Medchemexpress LLC SDZ 220-581 (hydrochloride) | 179411-93-9 | 50 MG
SDP
Catalog No. 5000338839
Encompass_Preferred
SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with a pKᵢ value of 7.7. It protects mice against maximal electroshock seizures (MES) in a dose-dependent manner, exhibiting a rapid onset and long duration of action upon oral administration. It targets iGluR, a membrane transporter/ion channel involved in neuronal signaling.
  • Mechanism of action: Competitive NMDA receptor antagonist.
  • Key features: Orally active, potent.
  • Target: iGluR (NMDA receptor).
  • Research application: Protection against maximal electroshock seizures (MES).
Catalog No. 50-003-38839 Supplier Medchemexpress LLC Supplier No. HY13059B50MG
SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with a pKᵢ value of 7.7. It protects mice against maximal electroshock seizures (MES) in a dose-dependent manner, exhibiting a rapid onset and long duration of action upon oral administration. It targets iGluR, a membrane transporter/ion channel involved in neuronal signaling.
  • Mechanism of action: Competitive NMDA receptor antagonist.
  • Key features: Orally active, potent.
  • Target: iGluR (NMDA receptor).
  • Research application: Protection against maximal electroshock seizures (MES).