Learn More
Medchemexpress LLC Sdz 220-581 hydrochloride | 179411-93-9 | 99.48% | 10MM/1ML
SDP
Catalog No. 5000341090
Encompass_Preferred
SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with a pKi value of 7.7. In animal studies, SDZ 220-581 dose-dependently protected mice against maximal electroshock seizures (MES) with a rapid onset and long duration of action.
  • Orally active
  • Potent NMDA receptor antagonist
  • Competitive NMDA receptor antagonist
  • Protects against maximal electroshock seizures
  • Rapid onset of action
  • Long duration of action
Catalog No. 50-003-41090 Supplier Medchemexpress LLC Supplier No. HY13059B10MM/1ML
SDZ 220-581 hydrochloride is an orally active, potent, competitive NMDA receptor antagonist with a pKi value of 7.7. In animal studies, SDZ 220-581 dose-dependently protected mice against maximal electroshock seizures (MES) with a rapid onset and long duration of action.
  • Orally active
  • Potent NMDA receptor antagonist
  • Competitive NMDA receptor antagonist
  • Protects against maximal electroshock seizures
  • Rapid onset of action
  • Long duration of action