Learn More
TARGETMOL CHEMICALS INC SDZ-62-434 1MG
SDP
Catalog No. 502879439
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. SDZ-62-434, a platelet-activating factor inhibitor with antitumor activity, inhibits bombesin- and platelet-derived growth factor-induced DNA synthesis in quiescent Swiss 3T3 cells, making it a potential candidate for studying leukemia, human xenograft colorectal cancer, and lung cancer. Purity 98.61%
Catalog No. 50-287-9439 Supplier TARGETMOL CHEMICALS INC Supplier No. T287401MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. SDZ-62-434, a platelet-activating factor inhibitor with antitumor activity, inhibits bombesin- and platelet-derived growth factor-induced DNA synthesis in quiescent Swiss 3T3 cells, making it a potential candidate for studying leukemia, human xenograft colorectal cancer, and lung cancer. Purity 98.61%