Learn More
Medchemexpress LLC HY-100164 10mg Medchemexpress, SDZ-MKS 492 CAS:114606-56-3 Purity:>98%
SDP
Catalog No. 501967713
Encompass_Preferred
Medchemexpress, HY-100164 10mg SDZ-MKS 492 CAS:114606-56-3 SDZ-MKS 492 (MKS 492) is a selective type III isozyme inhibitor of cyclic nucleotide phosphodiesterase, effective in allergic bronchoconstriction and platelet activating factor (PAF) or LTB4-induced inflammatory reactions in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7713 Supplier Medchemexpress LLC Supplier No. HY10016410MG
Medchemexpress, HY-100164 10mg SDZ-MKS 492 CAS:114606-56-3 SDZ-MKS 492 (MKS 492) is a selective type III isozyme inhibitor of cyclic nucleotide phosphodiesterase, effective in allergic bronchoconstriction and platelet activating factor (PAF) or LTB4-induced inflammatory reactions in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.