Learn More
Medchemexpress LLC Sdz-mks 492 | 114606-56-3 | MFCD00898843 | 99.8% | 433.46 g/mol | C20H27N5O6 | 1 ML
SDP
Catalog No. 5000215883
Encompass_Preferred
SDZ-MKS 492 is a selective cyclic GMP-inhibited phosphodiesterase (PDE III) inhibitor used in biochemical and cellular research to probe PDE-mediated signaling. It is supplied as research-grade solid material and as a ready-to-use 10 mM solution in DMSO for convenient dosing in assays.
  • Selective inhibition of cyclic GMP-inhibited phosphodiesterase (type III)
  • Available as 10 mM solution in DMSO and multiple solid sizes
  • High purity suitable for research applications
  • Characterized by CAS number and molecular data for reproducibility
  • Convenient ready-to-use stock reduces preparation time
Catalog No. 50-002-15883 Supplier Medchemexpress LLC Supplier No. HY10016410MM 1ML
SDZ-MKS 492 is a selective cyclic GMP-inhibited phosphodiesterase (PDE III) inhibitor used in biochemical and cellular research to probe PDE-mediated signaling. It is supplied as research-grade solid material and as a ready-to-use 10 mM solution in DMSO for convenient dosing in assays.
  • Selective inhibition of cyclic GMP-inhibited phosphodiesterase (type III)
  • Available as 10 mM solution in DMSO and multiple solid sizes
  • High purity suitable for research applications
  • Characterized by CAS number and molecular data for reproducibility
  • Convenient ready-to-use stock reduces preparation time