Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SDZ281-977 (SDZ-LAP 977) | 150779-71-8 | MFCD00920170 | 98.0% | 316.35 | C18H20O5 | 50 MG
SDP

Catalog No. 5000245381
Encompass_Preferred

SDZ281-977 is a small-molecule derivative of Lavendustin A with inhibitory activity against the epidermal growth factor receptor (EGFR) tyrosine kinase. Supplied as a solid for research use only, the compound is suitable for biochemical, cellular, and in vivo studies of EGFR signaling and antiproliferative effects.

  • Derivative of Lavendustin A with EGFR inhibitory activity.
  • High purity (98.0%).
  • Molecular weight 316.35; formula C18H20O5.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be required.
  • In vivo formulations documented for PEG300/Tween-80, SBE-β-CD, and corn oil.
  • Stable as powder at -20°C (up to 3 years) and 4°C (up to 2 years).

Catalog No. 50-002-45381 Supplier Medchemexpress LLC Supplier No. HY10175650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SDZ281-977 is a small-molecule derivative of Lavendustin A with inhibitory activity against the epidermal growth factor receptor (EGFR) tyrosine kinase. Supplied as a solid for research use only, the compound is suitable for biochemical, cellular, and in vivo studies of EGFR signaling and antiproliferative effects.

  • Derivative of Lavendustin A with EGFR inhibitory activity.
  • High purity (98.0%).
  • Molecular weight 316.35; formula C18H20O5.
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance may be required.
  • In vivo formulations documented for PEG300/Tween-80, SBE-β-CD, and corn oil.
  • Stable as powder at -20°C (up to 3 years) and 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.