Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Se-[(2,2-dimethyl-1,3-dioxolan-4-yl)methyl] 4-chlorobenzoselenolate | 2098850-81-6 | 98.9% | C13H15ClO3Se | 10MG
SDP

Supplier:  Medchemexpress LLC HY13970310MG

Encompass_Preferred

Se-DMC is a selenium-containing small-molecule research compound reported to have anti-inflammatory effects in preclinical studies. In a mouse complete Freund's adjuvant (CFA) model it attenuated inflammatory responses, nociception, and related neurobehavioral deficits. The compound is supplied as a high-purity reagent with specified solubility and storage recommendations for laboratory use.

  • High purity (98.9%).
  • Molecular formula C13H15ClO3Se; molecular weight 333.67 g·mol⁻1.
  • Reported activity in mouse CFA model of inflammation.
  • Soluble in DMSO (50 mg/mL); ultrasonic assistance recommended.
  • Storage under nitrogen recommended; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Intended for biochemical assays and inflammation research.

Catalog No. 50-002-61446


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.