Learn More
Lonza Walkersville SE Cell Line 4D-Nucleofector X Kit L
Catalog No. NC1483148
Encompass
The SE Cell Line 4D-Nucleofector X Kit L is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput applicationsContent:2 x 0.675 mL SE Cell Line Nucleofector Solution2 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)12 x 100 µL Nucleocuvette Vessel12 x Single use pipettes
Catalog No. NC1483148 Supplier Lonza Walkersville Supplier No. V4XC1012
The SE Cell Line 4D-Nucleofector X Kit L is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput applicationsContent:2 x 0.675 mL SE Cell Line Nucleofector Solution2 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)12 x 100 µL Nucleocuvette Vessel12 x Single use pipettes