Learn More
Lonza Walkersville SE Cell Line 4D-Nucleofector X Kit L
Catalog No. NC0475784
Encompass
The SE Cell Line 4D-Nucleofector X Kit L is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System (including 4D-Nucleofector including LV Unit), 96-well Shuttle System and 384-well Nucleofector System Applications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput applicationsContent:2.25 mL SE Cell Line Nucleofector Solution0.5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)24 x 100 µL Nucleocuvette Vessel,24 x Single use pipettes
Catalog No. NC0475784 Supplier Lonza Walkersville Supplier No. V4XC1024
The SE Cell Line 4D-Nucleofector X Kit L is one of our three 4D-Nucleofector Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System (including 4D-Nucleofector including LV Unit), 96-well Shuttle System and 384-well Nucleofector System Applications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput applicationsContent:2.25 mL SE Cell Line Nucleofector Solution0.5 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)24 x 100 µL Nucleocuvette Vessel,24 x Single use pipettes