Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Se-Methylselenocyste 10mM 1mL | 26046-90-2 | 1 ML
SDP

Catalog No. 5000229667
Encompass_Preferred

Se-methylselenocysteine is an organoselenium amino-acid derivative used as a biochemical reagent and research compound. The manufacturer provides physicochemical data, solubility guidance for preparing aqueous stock solutions, and storage recommendations.

  • Organoselenium amino acid (CAS 26046-90-2).
  • Molecular weight 182.08 g/mol; formula C4H9NO2Se.
  • Purity 99.6%.
  • Solubility: water 62.5 mg/mL (≈343.3 mM) with ultrasonic assistance; DMSO <1 mg/mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Datasheet includes preparations for 1 mM, 5 mM, and 10 mM aqueous stock solutions from the solid.

Catalog No. 50-002-29667 Supplier Medchemexpress LLC Supplier No. HY11424510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Se-methylselenocysteine is an organoselenium amino-acid derivative used as a biochemical reagent and research compound. The manufacturer provides physicochemical data, solubility guidance for preparing aqueous stock solutions, and storage recommendations.

  • Organoselenium amino acid (CAS 26046-90-2).
  • Molecular weight 182.08 g/mol; formula C4H9NO2Se.
  • Purity 99.6%.
  • Solubility: water 62.5 mg/mL (≈343.3 mM) with ultrasonic assistance; DMSO <1 mg/mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Datasheet includes preparations for 1 mM, 5 mM, and 10 mM aqueous stock solutions from the solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.