Learn More
Apexbio Technology LLC Se-Methylselenocysteine 26046-90-2 25mg
SDP
Supplier:  Apexbio Technology LLC A463125
Encompass_Preferred
Se-Methylselenocysteine (CAS 26046-90-2) is an organoselenium compound that serves as a selenium donor and is investigated for its chemopreventive properties It is metabolized in vivo to methylselenol which is believed to modulate cellular redox status and influence apoptotic pathways Research has focused on its ability to inhibit tumor development particularly in prostate carcinoma models by affecting cellular proliferation and promoting apoptosis Se-Methylselenocysteine is utilized in studies investigating the prevention of neoplastic progression and disease recurrence with applications in oncology and selenium metabolism research
Catalog No. 50-000-93952