Learn More
Medchemexpress LLC HY-114245 5mg Medchemexpress, Se-Methylselenocysteine CAS:26046-90-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1142455MG
Encompass_Preferred
Medchemexpress, HY-114245 5mg Se-Methylselenocysteine CAS:26046-90-2 Se-Methylselenocysteine, a precursor of Methylselenol, has potent cancer chemopreventive activity and anti-oxidant activity. Se-Methylselenocysteine is orally bioavailable, and induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9313