Learn More
Cpi International SE STD CA 250ML
SDP
Catalog No. NC1190208
Encompass
CPI International's Peak Performance Calcium (Ca) standard is provided at a concentration of 10,000 ppm and a volume of 250 mL. 10,000 ug/mL. In 4% HNO3 Matrix - 18 month shelf-life.
Catalog No. NC1190208 Supplier Cpi International Supplier No. 440010M91
Add to Cart
Add to Cart
CPI International's Peak Performance Calcium (Ca) standard is provided at a concentration of 10,000 ppm and a volume of 250 mL. 10,000 ug/mL. In 4% HNO3 Matrix - 18 month shelf-life.