Learn More
Cpi International Peak Performance Inorganic Standard, Chromium (Cr), 10000 μg/mL
SDP
Catalog No. NC1159100
Encompass
Peak Performance Inorganic Single-Element Standard.Chromium (Cr).10,000 ug/mL.In 4% HNO3 Matrix.100 mL.Guaranteed accuracy to +/- 0.3%.ISO 17025 accredited and 9001 certified.18 month shelf-life.
Catalog No. NC1159100 Supplier Cpi International Supplier No. 440010M121
Add to Cart
Add to Cart
Peak Performance Inorganic Single-Element Standard.Chromium (Cr).10,000 ug/mL.In 4% HNO3 Matrix.100 mL.Guaranteed accuracy to +/- 0.3%.ISO 17025 accredited and 9001 certified.18 month shelf-life.