Learn More
Cpi International SE STD RHODIUM (RH) 250ML
SDP
Catalog No. NC1305031
Encompass
SE Std Rhodium (Rh), 2% to 10% HCl, 1000ug/mL, 250mL
Catalog No. NC1305031 Supplier Cpi International Supplier No. 44001000442
SE Std Rhodium (Rh), 2% to 10% HCl, 1000ug/mL, 250mL