Learn More
Hoefer, Inc SE300 miniVE Integrated Vertical Protein Electrophoresis (Basic unit)
SDP
Supplier:  Hoefer, Inc SE300
Encompass_Preferred
Perform both electrophoresis and electro transfer in one sturdy, compact unit. Features:Quick, excellent results using minimal bufferPassive cooling produces gels with straight lanes and sharp bandsCast your own or use precast gels from a variety of manufacturersTransfer up to 4 gels in 45 minutes with only 300 ml buffer using optional SE302 Blot Modules Designed for use with a wide variety of precast gelsComplete Unit Includes:Buffer Chamber; Gel Modules with Gaskets, 2 assemblies; Safety Lid with High Voltage Leads; Glass Plates, Rectangular, 10 x 10.5 cm, 3 pcs; Glass Plates, Notched, 10 x 10.5 cm, 3 pcs; Combs, 10-well, 1.0 mm thick, 2 pcs; T-Spacers, 1.0 mm thick, 4 pcs;Basic Unit Includes:The same as complete unit without the combs, spacers, and glass plates. Specifications:Glass Plate Size (W x H) 10 x 10.5 cm or 10 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 1000 V, 500 mA, 50 WUnit Dimensions (W x H x D) 32 x 37 x 14 cm
Catalog No. NC2956968