Learn More
Azzota Corp SE3000 UV-Vis spectro 4nm
Catalog No. 5000887310
Encompass_Preferred
SE3000 single beam UV-Vis scanning spectrophotometer 4nm bandwidth 190-1100nm 320 x 240 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function
Catalog No. 50-008-87310 Supplier Azzota Corp Supplier No. SE3000
SE3000 single beam UV-Vis scanning spectrophotometer 4nm bandwidth 190-1100nm 320 x 240 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function