Learn More
Azzota Corp SE3300 UV-Vis spectro 1nm
Catalog No. 5000887290
Encompass_Preferred
SE3300 single beam UV-Vis scanning spectrophotometer 1nm bandwidth 190-1100nm 320 x 240 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function
Catalog No. 50-008-87290 Supplier Azzota Corp Supplier No. SE3300
SE3300 single beam UV-Vis scanning spectrophotometer 1nm bandwidth 190-1100nm 320 x 240 LCD Automatically set WL USB connection Can display and save 200 standard curves and 200 groups of data With Kinetics Function