Learn More
Hoefer, Inc Hoefer™ SE600X Chroma Deluxe Dual Cooled Vertical Protein Electrophoresis Unit (Complete unit)
SDP
Catalog No. NC2740655
Encompass_Preferred
Ergonomic design offers easier handling and sample introductionFeatures:Maximum glass plate size 18 x 16 cmGels are fully submerged for greater temperature equilibrationProduces straight lanes and sharp,well-defined bandsRun up to four gels (maximum 112 samples) at one time under identical conditionsAccommodates denaturing and native polyacrylamide gels, and the second-dimension of 2-D electrophoresisRun gels at uniform temperature from 1 to 45°CA wide array of accessories enables you to tailor gel configurations to your needsComplete Unit Includes:Buffer Chamber; Safety Lid w/High Voltage Leads; Heat Exchanger; Glass Plates 18 x 16 cm-6 pcs; Combs, 15-well, 1.5 mm thick, 2 pcs; Spacers, 1.5 mm thick, 2 cm wide, 4 pcs; Dual Gel CasterSpecifications:Glass Plate Size (W x H) 18 x 16 cm or 18 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 1000 V, 500 mA, 50 WMaximum Temperature 45°CUnit Dimensions (W x H x D) 32 x 29 x 14 cm
Catalog No. NC2740655 Supplier Hoefer, Inc Supplier No. SE600X151.5
Ergonomic design offers easier handling and sample introductionFeatures:Maximum glass plate size 18 x 16 cmGels are fully submerged for greater temperature equilibrationProduces straight lanes and sharp,well-defined bandsRun up to four gels (maximum 112 samples) at one time under identical conditionsAccommodates denaturing and native polyacrylamide gels, and the second-dimension of 2-D electrophoresisRun gels at uniform temperature from 1 to 45°CA wide array of accessories enables you to tailor gel configurations to your needsComplete Unit Includes:Buffer Chamber; Safety Lid w/High Voltage Leads; Heat Exchanger; Glass Plates 18 x 16 cm-6 pcs; Combs, 15-well, 1.5 mm thick, 2 pcs; Spacers, 1.5 mm thick, 2 cm wide, 4 pcs; Dual Gel CasterSpecifications:Glass Plate Size (W x H) 18 x 16 cm or 18 x 8 cmSpacers 0.75 mm, 1.0 mm, 1.5 mmMaximum Power Settings 1000 V, 500 mA, 50 WMaximum Temperature 45°CUnit Dimensions (W x H x D) 32 x 29 x 14 cm