Learn More
Azzota Corp SE6100 double beam spectro
Catalog No. 5000887281
Encompass_Preferred
SE6100 Double beam scanning spectrophotometer large screen high accuracy Wavelength 190-1100nm bandwidth 1 8nm
Catalog No. 50-008-87281 Supplier Azzota Corp Supplier No. SE6100
SE6100 Double beam scanning spectrophotometer large screen high accuracy Wavelength 190-1100nm bandwidth 1 8nm