Learn More
Chondrex Inc SEA DETECTION KIT, 1 KT
Catalog No. 501526786
Encompass_Preferred
Assay kit to determine Staphylococcal Enterotoxin A (SEA) levels in food, feces, intestinal fluids and other liquid samples. Kit comes with a 96-well ELISA plate (pre-coated with capture antibody) and all reagents necessary to complete the assays. Standard Range: 10 ng/mL to 0.6 ng/mL. Working Time: 1.5 Hours.
Catalog No. 50-152-6786 Supplier Chondrex Inc Supplier No. 6029
Add to Cart
Add to Cart
Assay kit to determine Staphylococcal Enterotoxin A (SEA) levels in food, feces, intestinal fluids and other liquid samples. Kit comes with a 96-well ELISA plate (pre-coated with capture antibody) and all reagents necessary to complete the assays. Standard Range: 10 ng/mL to 0.6 ng/mL. Working Time: 1.5 Hours.