Learn More
Lake Products Co SEA-SALT (ASTM D-1141-98) 17OZ
SDP
Catalog No. NC0548778
Encompass
"Sea Salt" ASTM D1141-98 - 17oz Bottle - One 17oz bottle makes approximately 3 gallons of artificial seawater.
Catalog No. NC0548778 Supplier Lake Products Co Supplier No. 11410317OZ.
"Sea Salt" ASTM D1141-98 - 17oz Bottle - One 17oz bottle makes approximately 3 gallons of artificial seawater.