Learn More
Analytical Sales & Services Vortexer
Catalog No. 50646501
Encompass
Sea Star Vortexer; Speed Range: Variabe 500-3000rpm (shaking or vortexing); 3 Position Switch; Orbit: 4.2mm; 90-110V AC, 50/60 Hz
Catalog No. 50-646-501 Supplier Analytical Sales & Services Supplier No. 6100800
Sea Star Vortexer; Speed Range: Variabe 500-3000rpm (shaking or vortexing); 3 Position Switch; Orbit: 4.2mm; 90-110V AC, 50/60 Hz