Learn More
3B Scientific™ Sea Urchin Embryology Slide Set
SDPSCI_ED
Catalog No. S95064L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
12
Catalog No. Quantity
S95064L 12
1 options
Catalog No. S95064L Supplier 3B Scientific™ Supplier No. W13055
Add to Cart
Add to Cart
Psammechinus miliaris
12 microscope slides:
  • 1(d). Sea urchin, unfertilized eggs
  • 2(d). Sea urchin, fertilized eggs
  • 3(d). Sea urchin, two cells
  • 4(d). Sea urchin, four cells
  • 5(d). Sea urchin, eight cells
  • 6(d). Sea urchin, sixteen cells
  • 7(d). Sea urchin, thirty-two cells
  • 8(d). Sea urchin, morula
  • 9(d). Sea urchin, blastula
  • 10(d). Sea urchin, blastula, beginning gastrulation
  • 11(d). Sea urchin, blastula, progressive gastrulation
  • 12(d). Sea urchin, pluteus larva
Specifications
Type Sea Urchin
Quantity 12