Learn More
Enterprise Technology Solutions Seagate 1TB One Touch USB 3.2 Gen 1 External Hard Drive with Password Protection (Black)
SDP
Catalog No. 5000172993
Encompass_Preferred
Protected by password-activated AES-256 hardware encryption, the 1TB One Touch USB 3.2 Gen 1 External Hard Drive from Seagate lets you store and access photos and files on the go. Beneath its black exterior, the One Touch is equipped with a 1TB hard drive and a bus-powered micro-USB 3.2 Gen 1 (USB 3.1 Gen 1/3.0) interface. The One Touch is formatted as exFAT, meaning it will work with most Windows and Mac systems
Catalog No. 50-001-72993 Supplier Enterprise Technology Solutions Supplier No. STKY1000400
Protected by password-activated AES-256 hardware encryption, the 1TB One Touch USB 3.2 Gen 1 External Hard Drive from Seagate lets you store and access photos and files on the go. Beneath its black exterior, the One Touch is equipped with a 1TB hard drive and a bus-powered micro-USB 3.2 Gen 1 (USB 3.1 Gen 1/3.0) interface. The One Touch is formatted as exFAT, meaning it will work with most Windows and Mac systems