Learn More
Agilent Technologies SEAHORSE XF GLUTAMINE OXIDATIO
Catalog No. NC1841740
Encompass
Seahorse XF Glutamine Oxidation Stress Test Kit. 3 Pouches per kit. Each pouch contains 1 each of BPTES, Oligomycin, FCCP and Rotenone/Antimycin A
Catalog No. NC1841740 Supplier Agilent Technologies Supplier No. 103674100
Add to Cart
Add to Cart
Seahorse XF Glutamine Oxidation Stress Test Kit. 3 Pouches per kit. Each pouch contains 1 each of BPTES, Oligomycin, FCCP and Rotenone/Antimycin A