Learn More
Agilent Technologies Seahorse XF Glycolytic Rate Assay Kit
Catalog No. NC1762049
Encompass
Seahorse XF Glycolytic Rate Assay Kit. Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of rotenone/antimycin A and 2-deoxy-D-glucose. For use with XFe/XF96 and XFe24 analyzers. Not compatible with XF24 and XF24-3 analyzers.
Catalog No. NC1762049 Supplier Agilent Technologies Supplier No. 103344100
Seahorse XF Glycolytic Rate Assay Kit. Each kit contains six (6) single-use pouches. Each pouch contains one (1) each of rotenone/antimycin A and 2-deoxy-D-glucose. For use with XFe/XF96 and XFe24 analyzers. Not compatible with XF24 and XF24-3 analyzers.